3-Fluoro-5-iodo-4-methoxybenzoic acid 95%

Supplier: Apollo Scientific
Warning

PC56064-1G PC56064-250MG
APOSPC56064-1GEA 568 EUR
APOSPC56064-1G APOSPC56064-250MG
3-Fluoro-5-iodo-4-methoxybenzoic acid 95%
3-Fluoro-5-iodo-4-methoxybenzoic acid
Formula: C₈H₆FIO₃
Storage Temperature: Refrigerator
MDL Number: MFCD24107165
CAS Number: 1542632-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR