Methyl 4-bromo-3-(difluoromethoxy)benzoate

Supplier: Apollo Scientific
Warning

PC56116-1G PC56116-250MG PC56116-5G
APOSPC56116-1GEA 36.4 EUR
APOSPC56116-1G APOSPC56116-250MG APOSPC56116-5G
Methyl 4-bromo-3-(difluoromethoxy)benzoate
Methyl 4-bromo-3-(difluoromethoxy)benzoate
Formula: C₉H₇BrF₂O₃
MW: 281,051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28401975
CAS Number: 1331943-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR