Potassium trans-2-methylcyclohexyltrifluoroborate
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Potassium trifluoro(trans-2-methylcyclohexyl)borate, Potassium, trifluoro-[(1R,2R)-2-methylcyclohexyl]boranuide
PC56185-5G
PC56185-1G
APOSPC56185-5GEA
735
EUR
APOSPC56185-5G
APOSPC56185-1G
Potassium trans-2-methylcyclohexyltrifluoroborate
Potassium trans-2-methylcyclohexyltrifluoroborate
Racemic
Formula:
C₇H₁₃BF₃·K MW: 204,083 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28955335 CAS Number: 1041642-14-1 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...