4-Bromo-3,5-difluorobenzaldehyde
Supplier: Apollo Scientific
|
Warning
|
PC56208-1G
PC56208-5G
PC56208-10G
APOSPC56208-1GEA
101
EUR
APOSPC56208-1G
APOSPC56208-5G
APOSPC56208-10G
4-Bromo-3,5-difluorobenzaldehyde
4-Bromo-3,5-difluorobenzaldehyde
Formula:
C₇H₃BrF₂O MW: 220,999 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16659085 CAS Number: 135564-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...