3,3,4,4,5,5,6,6-Octafluoroazepane-2,7-diimine

Supplier: Apollo Scientific
Warning

PC56221-1G PC56221-250MG
APOSPC56221-1GEA 400 EUR
APOSPC56221-1G APOSPC56221-250MG
3,3,4,4,5,5,6,6-Octafluoroazepane-2,7-diimine
3,3,4,4,5,5,6,6-Octafluoroazepane-2,7-diimine
Formula: C₆H₃F₈N₃
MW: 269.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30062895
CAS Number: 1980063-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR