2,3,5,6-Tetrafluorophenyl acrylate 97%

Supplier: Apollo Scientific
Warning

Synonyms: (2,3,5,6-Tetrafluorophenyl) prop-2-enoate

PC56225-5G PC56225-1G PC56225-25G
APOSPC56225-5GEA 405 EUR
APOSPC56225-5G APOSPC56225-1G APOSPC56225-25G
2,3,5,6-Tetrafluorophenyl acrylate 97%
2,3,5,6-Tetrafluorophenyl acrylate
Formula: C₉H₄F₄O₂
MW: 220,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129957
CAS Number: 101156-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR