2-Chloro-5-nitro-3-(trifluoromethyl)pyridine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-5-nitro-3-(trifluoromethyl)pyridine

PC56251-25G PC56251-5G
APOSPC56251-25GEA 177 EUR
APOSPC56251-25G APOSPC56251-5G
2-Chloro-5-nitro-3-(trifluoromethyl)pyridine 97%
2-Chloro-5-nitro-3-(trifluoromethyl)pyridine
Formula: C₆H₂ClF₃N₂O₂
MW: 226,54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00792435
CAS Number: 99368-67-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR