1-(2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl)-2,2,2-trifluoroethan-1-one 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1-(2-bromo-6-fluoro-3-methylphenyl)-2,2,2-trifluoroethanone

PC56255-1G PC56255-250MG
APOSPC56255-1GEA 655 EUR
APOSPC56255-1G APOSPC56255-250MG
1-(2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl)-2,2,2-trifluoroethan-1-one 95%
1-(2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl)-2,2,2-trifluoroethan-1-one
Formula: C₉H₅BrF₄O
MW: 285,033 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28141542
CAS Number: 1515433-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR