2-Fluoro-6-methyl-4-nitroaniline 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Fluoro-6-methyl-4-nitroaniline
PC56269-1G
PC56269-250MG
PC56269-5G
APOSPC56269-1GEA
524
EUR
APOSPC56269-1G
APOSPC56269-250MG
APOSPC56269-5G
2-Fluoro-6-methyl-4-nitroaniline 95%
2-Fluoro-6-methyl-4-nitroaniline
Formula:
C₇H₇FN₂O₂ MW: 170,141 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28130101 CAS Number: 1426805-91-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...