1,2,3-Trifluoro-4-iodo-5-(trifluoromethyl)benzene 95%

Supplier: Apollo Scientific
Warning

PC56677-1G PC56677-250MG
APOSPC56677-1GEA 522 EUR
APOSPC56677-1G APOSPC56677-250MG
1,2,3-Trifluoro-4-iodo-5-(trifluoromethyl)benzene 95%
1,2,3-Trifluoro-4-iodo-5-(trifluoromethyl)benzene
Formula: C₇HF₆I
Storage Temperature: Ambient
MDL Number: MFCD31613611
CAS Number: 2237235-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR