3-Bromo-5-(trifluoromethyl)phenylacetylene 95%

Supplier: Apollo Scientific
Warning

PC56842-1G PC56842-250MG
APOSPC56842-1GEA 102 EUR
APOSPC56842-1G APOSPC56842-250MG
3-Bromo-5-(trifluoromethyl)phenylacetylene 95%
3-Bromo-5-(trifluoromethyl)phenylacetylene
Formula: C₉H₄BrF₃
MW: 249.027 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21243959
CAS Number: 1494947-64-6
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR