3-Bromo-7-fluoro-2-(hydroxymethyl)benzo[b]thiophene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(3-Bromo-7-fluoro-1-benzothiophen-2-yl)methanol
PC56847-1G
PC56847-250MG
APOSPC56847-1GEA
654
EUR
APOSPC56847-1G
APOSPC56847-250MG
3-Bromo-7-fluoro-2-(hydroxymethyl)benzo[b]thiophene
3-Bromo-7-fluoro-2-(hydroxymethyl)benzo[b]thiophene
Formula:
C₉H₆BrFOS MW: 261,111 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29430045 CAS Number: 1569087-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...