6-Amino-2-fluoro-3-iodophenol 95%

Supplier: Apollo Scientific
Warning

PC56884-1G PC56884-250MG
APOSPC56884-1GEA 823 EUR
APOSPC56884-1G APOSPC56884-250MG
6-Amino-2-fluoro-3-iodophenol 95%
6-Amino-2-fluoro-3-iodophenol
Formula: C₆H₅FINO
MW: 253,013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31664913
CAS Number: 2090794-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR