5-Bromo-7-fluorobenzofuran-2-carbonyl chloride

Supplier: Apollo Scientific
Danger

PC56922-1G PC56922-250MG
APOSPC56922-1GEA 102 EUR
APOSPC56922-1G APOSPC56922-250MG
5-Bromo-7-fluorobenzofuran-2-carbonyl chloride
5-Bromo-7-fluorobenzofuran-2-carbonyl chloride
Formula: C₉H₃BrClFO₂
MW: 277,474 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31695607
CAS Number: 2379918-48-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR