2-(4-Amino-3-bromophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%
Supplier: Apollo Scientific
|
Warning
|
PC56937-1G
PC56937-250MG
APOSPC56937-1GEA
654
EUR
APOSPC56937-1G
APOSPC56937-250MG
2-(4-Amino-3-bromophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 95%
2-(4-Amino-3-bromophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Formula:
C₉H₆BrF₆NO MW: 338,044 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31695609 CAS Number: 637347-75-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...