Methyl 6-bromo-5-fluoropyridine-2-carboxylate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-3-fluoro-6-(methoxycarbonyl)pyridine, Methyl 6-bromo-5-fluoropicolinate
PC57015-1G
PC57015-250MG
APOSPC57015-1GEA
360
EUR
APOSPC57015-1G
APOSPC57015-250MG
Methyl 6-bromo-5-fluoropyridine-2-carboxylate
Methyl 6-bromo-5-fluoropyridine-2-carboxylate
Formula:
C₇H₅BrFNO₂ MW: 234,023 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14698111 CAS Number: 1210419-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...