2-Fluoro-7-chlorophenazine 95%
Supplier: Apollo Scientific
|
Warning
|
PC57079-1G
PC57079-250MG
APOSPC57079-1GEA
111
EUR
APOSPC57079-1G
APOSPC57079-250MG
2-Fluoro-7-chlorophenazine 95%
2-Fluoro-7-chlorophenazine
Formula:
C₁₂H₆ClFN₂ MW: 232.641 g/mol Melting Pt: 225…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD31922521 CAS Number: 1346682-89-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...