4-Fluoro-N-(2,2,2-trifluoroethyl)aniline
Supplier: Apollo Scientific
|
Warning
|
PC57091-1G
PC57091-250MG
APOSPC57091-1GEA
42.2
EUR
APOSPC57091-1G
APOSPC57091-250MG
4-Fluoro-N-(2,2,2-trifluoroethyl)aniline
4-Fluoro-N-(2,2,2-trifluoroethyl)aniline
Formula:
C₈H₇F₄N MW: 193,141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11153191 CAS Number: 62158-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...