2-Bromo-6-fluoro-3-iodopyridine 95%

Supplier: Apollo Scientific

PC57327-1G PC57327-250MG
APOSPC57327-1GEA 655 EUR
APOSPC57327-1G APOSPC57327-250MG
2-Bromo-6-fluoro-3-iodopyridine 95%
2-Bromo-6-fluoro-3-iodopyridine
Formula: C₅H₂BrFIN
MW: 301,883 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20923590
CAS Number: 1807231-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR