5-Bromo-2,3-difluorobenzeneboronic acid, pinacol ester 96%

Supplier: Apollo Scientific

Synonyms: 2-(5-Bromo-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC5752-100MG PC5752-500MG
APOSPC5752-100MGEA 219 EUR
APOSPC5752-100MG APOSPC5752-500MG
5-Bromo-2,3-difluorobenzeneboronic acid, pinacol ester 96%
5-Bromo-2,3-difluorobenzeneboronic acid, pinacol ester
Formula: C₁₂H₁₄BBrF₂O₂
MW: 318,95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11855989
CAS Number: 1073339-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR