2-Bromo-1,1,1,3-tetrafluorobut-2-ene 97%

Supplier: Apollo Scientific
Warning

PC5803-1G PC5803-5G
APOSPC5803-1GEA 116 EUR
APOSPC5803-1G APOSPC5803-5G
2-Bromo-1,1,1,3-tetrafluorobut-2-ene 97%
2-Bromo-1,1,1,3-tetrafluorobut-2-ene
Formula: C₄H₃BrF₄
MW: 206.964 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08461628
CAS Number: 933668-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR