4-(2,2,2-Trifluoroethoxy)phenol
Supplier: Apollo Scientific
|
Danger
|
PC5859-1G
PC5859-5G
APOSPC5859-1GEA
234
EUR
APOSPC5859-1G
APOSPC5859-5G
4-(2,2,2-Trifluoroethoxy)phenol
4-(2,2,2-Trifluoroethoxy)phenol
Formula:
C₈H₇F₃O₂ MW: 192,135 g/mol Melting Pt: 60…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972983 CAS Number: 129560-99-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...