4-Bromo-3-(trifluoromethyl)benzyl alcohol 98%

Supplier: Apollo Scientific
Warning

Synonyms: (4-Bromo-3-(trifluoromethyl)phenyl)methanol, [4-Bromo-3-(trifluoromethyl)phenyl]methanol, 2-Bromo-5-(hydroxymethyl)benzotrifluoride

PC6024-1G PC6024-5G PC6024-25G
APOSPC6024-1GEA 49.3 EUR
APOSPC6024-1G APOSPC6024-5G APOSPC6024-25G
4-Bromo-3-(trifluoromethyl)benzyl alcohol 98%
4-Bromo-3-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆BrF₃O
MW: 255,032 g/mol
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD08059129
CAS Number: 957207-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR