Ethyl 3-[(4-fluorophenyl)amino]propanoate 97%
Supplier: Apollo Scientific
Synonyms:
N-(Ethoxycarbonyl)ethyl-4-fluoroaniline
PC6097-1G
PC6097-5G
APOSPC6097-1GEA
157
EUR
APOSPC6097-1G
APOSPC6097-5G
Ethyl 3-[(4-fluorophenyl)amino]propanoate 97%
Ethyl 3-[(4-fluorophenyl)amino]propanoate
Formula:
C₁₁H₁₄FNO₂ MW: 211,233 g/mol Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD10567166 CAS Number: 887574-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...