N-Ethylidene-2,2,2-trifluoroethylamine

Supplier: Apollo Scientific
Warning

PC6126-1G PC6126-5G
APOSPC6126-1GEA 139 EUR
APOSPC6126-1G APOSPC6126-5G
N-Ethylidene-2,2,2-trifluoroethylamine
N-Ethylidene-2,2,2-trifluoroethylamine
Formula: C₄H₆F₃N
MW: 125,092 g/mol
Boiling Pt: 73…74 °C
Density: 1,1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD07777214
CAS Number: 80395-37-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR