3'-Bromo-4'-(trifluoromethoxy)acetanilide 99%

Supplier: Apollo Scientific
Warning

Synonyms: N-[3-Bromo-4-(trifluoromethoxy)phenyl]acetamide, 4-Acetamido-2-bromo-α,α,α-trifluoroanisole

PC6257-1G PC6257-5G
APOSPC6257-1GEA 55.3 EUR
APOSPC6257-1G APOSPC6257-5G
3'-Bromo-4'-(trifluoromethoxy)acetanilide 99%
3'-Bromo-4'-(trifluoromethoxy)acetanilide
Formula: C₉H₇BrF₃NO₂
MW: 298,057 g/mol
Melting Pt: 86…87 °C
Storage Temperature: Ambient
MDL Number: MFCD13176598
CAS Number: 755030-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR