3,5-Difluorostyrene 99%

Supplier: Apollo Scientific
Warning

Synonyms: 1,3-Difluoro-5-ethenylbenzene, 1,3-Difluoro-5-vinylbenzene

PC6277-1G PC6277-5G
APOSPC6277-1GEA 111 EUR
APOSPC6277-1G APOSPC6277-5G
3,5-Difluorostyrene 99%
3,5-Difluorostyrene
Formula: C₈H₆F₂
Storage Temperature: Refrigerator
MDL Number: MFCD08461660
CAS Number: 182132-57-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR