N-[1-(2-Fluorophenyl)ethyl]methylamine 98+%

Supplier: Apollo Scientific
Warning

Synonyms: α,N-Dimethyl-2-fluorobenzylamine, 1-(2-Fluorophenyl)-N-methylethylamine

PC6296-1G PC6296-250MG
APOSPC6296-1GEA 430 EUR
APOSPC6296-1G APOSPC6296-250MG
N-[1-(2-Fluorophenyl)ethyl]methylamine 98+%
N-[1-(2-Fluorophenyl)ethyl]methylamine
Formula: C₉H₁₂FN
MW: 153,197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08461632
CAS Number: 926213-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR