4,4,4-Trifluorobutan-2-ol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Hydroxy-1,1,1-trifluorobutane
PC6395-1G
APOSPC6395-1GEA
301
EUR
APOSPC6395-1G
4,4,4-Trifluorobutan-2-ol 97%
4,4,4-Trifluorobutan-2-ol
Formula:
C₄H₇F₃O MW: 128,093 g/mol Boiling Pt: 96…98 °C Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD07784221 CAS Number: 101054-93-7 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...