Potassium bromo(difluoro)acetate 97+%

Supplier: Apollo Scientific
Warning

Synonyms: Potassium 2-bromo-2,2-difluoroethanoate, Bromo(difluoro)acetic acid potassium salt, Potassium 2-bromo-2,2-difluoroacetate

PC6410-25G PC6410-5G
APOSPC6410-25GEA 430 EUR
APOSPC6410-25G APOSPC6410-5G
Potassium bromo(difluoro)acetate 97+%
Potassium bromo(difluoro)acetate
Formula: C₂BrF₂O₂·K
MW: 213,019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784243
CAS Number: 87189-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR