Ethyl 3-hydroxy-3-methyl-4,4,4-trifluorobutanoate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 3-hydroxy-3-methyl-4,4,4-trifluorobutyrate
PC6444-5G
PC6444-1G
APOSPC6444-5GEA
306
EUR
APOSPC6444-5G
APOSPC6444-1G
Ethyl 3-hydroxy-3-methyl-4,4,4-trifluorobutanoate 97%
Ethyl 3-hydroxy-3-methyl-4,4,4-trifluorobutanoate
Formula:
C₇H₁₁F₃O₃ MW: 200.156 g/mol Boiling Pt: 176 °C Density: 1.221 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD07784252 CAS Number: 649-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...