1H,1H-Nonadecafluoro-3,6,9-trioxatridecan-1-ol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Hydroxy-1H,1H-nonadecafluoro-3,6,9-trioxatridecane, 2,2-Difluoro-2-{1,1,2,2-tetrafluoro-2-[1,1,2,2-tetrafluoro-2-(nonafluorobutoxy)ethoxy]ethoxy}ethan-1-ol
PC6716-100G
PC6716-25G
PC6716-5G
APOSPC6716-100GEA
357
EUR
APOSPC6716-100G
APOSPC6716-25G
APOSPC6716-5G
1H,1H-Nonadecafluoro-3,6,9-trioxatridecan-1-ol
1H,1H-Nonadecafluoro-3,6,9-trioxatridecan-1-ol
Formula:
C₁₀H₃F₁₉O₄ MW: 548,098 g/mol Boiling Pt: 175…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094406 CAS Number: 317817-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...