2,3,5-Trifluorochlorobenzene 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Chloro-2,3,5-trifluorobenzene

PC6817-1G PC6817-5G
APOSPC6817-1GEA 69.8 EUR
APOSPC6817-1G APOSPC6817-5G
2,3,5-Trifluorochlorobenzene 97%
2,3,5-Trifluorochlorobenzene
Formula: C₆H₂ClF₃
MW: 166,528 g/mol
Boiling Pt: 123 °C
Melting Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00070763
CAS Number: 2367-77-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR