2-Bromo-4-fluorophenacyl bromide
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Bromo-1-(2-bromo-4-fluorophenyl)ethan-1-one, 2,2'-Dibromo-4-fluoroacetophenone
PC6831-1G
PC6831-5G
APOSPC6831-1GEA
128
EUR
APOSPC6831-1G
APOSPC6831-5G
2-Bromo-4-fluorophenacyl bromide
2-Bromo-4-fluorophenacyl bromide
Formula:
C₈H₅Br₂FO MW: 295,931 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847026 CAS Number: 594810-90-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...