2-Bromo-3,3,3-trifluoropropyl acetate 97%
Supplier: Apollo Scientific
PC6832-10G
PC6832-1G
PC6832-5G
APOSPC6832-10GEA
524
EUR
APOSPC6832-10G
APOSPC6832-1G
APOSPC6832-5G
2-Bromo-3,3,3-trifluoropropyl acetate 97%
2-Bromo-3,3,3-trifluoropropyl acetate
Formula:
C₅H₆BrF₃O₂ Boiling Pt: 68 °C (100 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD09763643 CAS Number: 383-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...