1-[4-(Trifluoromethyl)phenyl]propylamine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-[4-(Trifluoromethyl)phenyl]propan-1-amine, 1-(4-(Trifluoromethyl)phenyl)propan-1-amine

PC6846-1G PC6846-250MG
APOSPC6846-1GEA 272 EUR
APOSPC6846-1G APOSPC6846-250MG
1-[4-(Trifluoromethyl)phenyl]propylamine 98%
1-[4-(Trifluoromethyl)phenyl]propylamine
Formula: C₁₀H₁₂F₃N
Boiling Pt: 79,5…80.5 °C (4 mm Hg)
Density: 1,1 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD07784290
CAS Number: 885276-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR