4-Chloro-8-(trifluoromethoxy)-2-(trifluoromethyl)quinoline

Supplier: Apollo Scientific
Warning

PC6847-100MG PC6847-1G
APOSPC6847-100MGEA 65.5 EUR
APOSPC6847-100MG APOSPC6847-1G
4-Chloro-8-(trifluoromethoxy)-2-(trifluoromethyl)quinoline
4-Chloro-8-(trifluoromethoxy)-2-(trifluoromethyl)quinoline
Formula: C₁₁H₄ClF₆NO
MW: 315,599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209304
CAS Number: 306935-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR