Bromofluoroacetic acid 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Bromofluoroacetic Acid, 2-Bromo-2-fluoroethanoic acid, Fluorobromoacetic acid
PC6862-1G
PC6862-5G
APOSPC6862-1GEA
133
EUR
APOSPC6862-1G
APOSPC6862-5G
Bromofluoroacetic acid 97%
Bromofluoroacetic acid
Formula:
C₂H₂BrFO₂ Boiling Pt: 98…99 °C (29 mm Hg) Melting Pt: 51…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD07784239 CAS Number: 359-25-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...