1H,1H,6H-Decafluorohexan-1-ol 98%

Supplier: Apollo Scientific
Warning

PC6969-1G PC6969-250MG PC6969-5G
APOSPC6969-1GEA 227 EUR
APOSPC6969-1G APOSPC6969-250MG APOSPC6969-5G
1H,1H,6H-Decafluorohexan-1-ol 98%
1H,1H,6H-Decafluorohexan-1-ol
Formula: C₆H₄F₁₀O
MW: 282,079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01025150
CAS Number: 60838-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR