(1E)-1H-Pentafluoroprop-1-ene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
E-HFC-1225ye, (1E)-1,2,3,3,3-Pentafluoroprop-1-ene
PC6994-25G
PC6994-5G
APOSPC6994-25GEA
1350
EUR
APOSPC6994-25G
APOSPC6994-5G
(1E)-1H-Pentafluoroprop-1-ene
(1E)-1H-Pentafluoroprop-1-ene
Formula:
C₃HF₅ MW: 132.032 g/mol Boiling Pt: –11…–9 °C Density: 1.336 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD02262202 CAS Number: 5595-10-8 UN: 3163 ADR: 2.2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...