Allyl chlorodifluoroacetate

Supplier: Apollo Scientific

PC7003-25G PC7003-5G
APOSPC7003-25GEA 321 EUR
APOSPC7003-25G APOSPC7003-5G
Allyl chlorodifluoroacetate
Allyl chlorodifluoroacetate
Formula: C₅H₅ClF₂O₂
MW: 170,542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033650
CAS Number: 118337-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR