3-Cyano-1,2-dihydro-1-(4-fluorophenyl)-2-oxopyridine

Supplier: Apollo Scientific

Synonyms: 1,2-Dihydro-1-(4-fluorophenyl)-2-oxopyridine-3-carbonitrile

PC7132-1G PC7132-250MG
APOSPC7132-1GEA 379 EUR
APOSPC7132-1G APOSPC7132-250MG
3-Cyano-1,2-dihydro-1-(4-fluorophenyl)-2-oxopyridine
3-Cyano-1,2-dihydro-1-(4-fluorophenyl)-2-oxopyridine
Formula: C₁₂H₇FN₂O
MW: 214,195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258781
CAS Number: 929000-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR