3-Bromo-5-fluorobenzal chloride 97%

Supplier: Apollo Scientific
Danger

PC7142-1G PC7142-5G
APOSPC7142-1GEA 105 EUR
APOSPC7142-1G APOSPC7142-5G
3-Bromo-5-fluorobenzal chloride 97%
3-Bromo-5-fluorobenzal chloride
Formula: C₇H₄BrCl₂F
MW: 257,915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258971
CAS Number: 1000339-58-1
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR