Chloro(fluoro)phenylacetonitrile 97%
Supplier: Apollo Scientific
|
Warning
|
PC7151-1G
PC7151-250MG
PC7151-5G
APOSPC7151-1GEA
270
EUR
APOSPC7151-1G
APOSPC7151-250MG
APOSPC7151-5G
Chloro(fluoro)phenylacetonitrile 97%
Chloro(fluoro)phenylacetonitrile
Formula:
C₈H₅ClFN MW: 169,583 g/mol Flash Pt: >90 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258967 CAS Number: 948014-31-1 UN: 2810 ADR: 6.1,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...