1-Fluoropropan-2-ol 97%
Supplier: Apollo Scientific
|
Warning
|
PC7229-1G
PC7229-5G
APOSPC7229-1GEA
183
EUR
APOSPC7229-1G
APOSPC7229-5G
1-Fluoropropan-2-ol 97%
1-Fluoropropan-2-ol
Formula:
C₃H₇FO Boiling Pt: 107…108 °C Density: 1,021 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00039620 CAS Number: 430-50-2 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...