1H,1H,2H,2H,3H,3H,4H,4H,5H,5H-Perfluoroundecyl acrylate 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6,6,7,7,8,8,9,9,10,10,11,11,11-Tridecafluoroundecyl acrylate
PC7254-1G
PC7254-5G
APOSPC7254-1GEA
139
EUR
APOSPC7254-1G
APOSPC7254-5G
1H,1H,2H,2H,3H,3H,4H,4H,5H,5H-Perfluoroundecyl acrylate 95%
1H,1H,2H,2H,3H,3H,4H,4H,5H,5H-Perfluoroundecyl acrylate
Formula:
C₁₄H₁₃F₁₃O₂ MW: 460,231 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09749660 CAS Number: 1000339-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...