6-Fluoro-2-methyl-1H-indole 98%

Supplier: Apollo Scientific

PC7443-1G PC7443-250MG
APOSPC7443-1GEA 184 EUR
APOSPC7443-1G APOSPC7443-250MG
6-Fluoro-2-methyl-1H-indole 98%
6-Fluoro-2-methyl-1H-indole
Formula: C₉H₈FN
MW: 149,165 g/mol
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD09027714
CAS Number: 40311-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR