4-Carboxy-2-fluorophenylboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Carboxy-2-fluorobenzeneboronic acid
PC7538-1G
PC7538-5G
APOSPC7538-1GEA
84.4
EUR
APOSPC7538-1G
APOSPC7538-5G
4-Carboxy-2-fluorophenylboronic acid 98%
4-Carboxy-2-fluorophenylboronic acid
Formula:
C₇H₆BFO₄ MW: 183,93 g/mol Melting Pt: 213…215 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08458473 CAS Number: 851335-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...