5-Bromo-4-chloro-2-fluorotoluene 97%

Supplier: Apollo Scientific
Warning

PC7758-1G PC7758-5G PC7758-25G
APOSPC7758-1GEA 21.9 EUR
APOSPC7758-1G APOSPC7758-5G APOSPC7758-25G
5-Bromo-4-chloro-2-fluorotoluene 97%
5-Bromo-4-chloro-2-fluorotoluene
Formula: C₇H₅BrClF
MW: 223,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143430
CAS Number: 201849-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR