2-Methyl-3,3,3-trifluoropropanal 95%

Supplier: Apollo Scientific
Warning

PC7762-1G PC7762-5G
APOSPC7762-1GEA 139 EUR
APOSPC7762-1G APOSPC7762-5G
2-Methyl-3,3,3-trifluoropropanal 95%
2-Methyl-3,3,3-trifluoropropanal
Formula: C₄H₅F₃O
MW: 126,077 g/mol
Boiling Pt: 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042425
CAS Number: 58928-28-2
UN: 1989
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR